Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001443.2 | NP_001434.1 | 1 | 87 | 82 | |
| GenBank mRNA | BC022287.1 | - | 1 | 72 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC022287.1, NM_001443.2 transcripts.
Set Membership: |
|

FABP1
Genomic Map