Assay Details
| Gene Symbol: SP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172712.1 | NP_001166183.1 | 2 | 78 | ||
| NM_003111.4 | NP_003102.1 | 2 | 78 | |||
| GenBank mRNA | AJ310752.1 | - | 1 | 52 | 78 | |
| AJ430844.1 | - | 1 | 52 | 78 | ||
| AK304199.1 | - | 2 | 78 | |||
| AW204666.1 | - | 2 | 78 | |||
| AY441957.1 | - | 2 | 78 | |||
| AY441958.1 | - | 1 | 78 | |||
| BC126414.1 | - | 2 | 8992130 | 78 | ||
| BC143928.1 | - | 2 | 9052446 | 78 | ||
More Information
Set Membership: |
|

SP3
Genomic Map