Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000509.5 | NP_000500.2 | 7 | 967 | 68 | |
| NM_021870.2 | NP_068656.2 | 7 | 909 | 68 | ||
| GenBank mRNA | AK289422.1 | - | 7 | 812 | 68 | |
| AK290824.1 | - | 7 | 858 | 68 | ||
| AL531173.3 | - | 3 | 68 | |||
| BC007044.1 | - | 7 | 814 | 3934943 | 68 | |
| BC021674.1 | - | 7 | 816 | 3935073 | 68 | |
| BT007081.1 | - | 7 | 768 | 68 | ||
| BX538098.1 | - | 6 | 656 | 68 | ||
| X00086.1 | - | 2 | 164 | 68 | ||
| X51473.1 | - | 5 | 546 | 68 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF118092.1 | - | Chr. - - [-] on Build | 68 | |||
More Information
Set Membership: |
|

FGG
Genomic Map