Assay Details
| Gene Symbol: HEXB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000521.3 | NP_000512.1 | 11 | 1452 | 106 | |
| NM_001292004.1 | NP_001278933.1 | 11 | 1572 | 106 | ||
| GenBank mRNA | AF378118.1 | - | 11 | 1407 | 106 | |
| AK122992.1 | - | 12 | 1309 | 106 | ||
| AK130002.1 | - | 11 | 1436 | 106 | ||
| AK130375.1 | - | 11 | 1363 | 106 | ||
| AY643499.1 | - | 11 | 1577 | 106 | ||
| BC017378.2 | - | 11 | 1357 | 2967035 | 106 | |
| BT009919.1 | - | 11 | 1335 | 106 | ||
| D10518.1 | - | 2 | 166 | 106 | ||
| FJ224336.1 | - | 11 | 1577 | 106 | ||
| GQ472222.1 | - | 11 | 1577 | 106 | ||
| M13519.1 | - | 11 | 1383 | 106 | ||
| M19735.1 | - | 11 | 1299 | 106 | ||
More Information
Set Membership: |
|

HEXB
Genomic Map