Assay Details
| Gene Symbol: RPS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017.2 | NP_001008.1 | 3 | 132 | 96 | |
| GenBank mRNA | AK312060.1 | - | 3 | 126 | 96 | |
| BC000475.1 | - | 1 | 111 | 2961637 | 96 | |
| BC006772.1 | - | 1 | 113 | 2899987 | 96 | |
| BC029732.1 | - | 3 | 112 | 4388273 | 96 | |
| BC066322.1 | - | 3 | 126 | 4816284 | 96 | |
| BC100032.1 | - | 3 | 112 | 6025884 | 96 | |
| L01124.1 | - | 3 | 132 | 96 | ||
| L05090.1 | - | 2 | 68 | 96 | ||
| X79239.1 | - | 3 | 100 | 96 | ||
More Information
Set Membership: |
|

RPS13
Genomic Map