Assay Details
Gene Symbol: ZNF391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076781.2 | NP_001070249.1 | 3 | 796 | 120 | |
NM_001322288.1 | NP_001309217.1 | 4 | 1020 | 120 | ||
NM_001322289.1 | NP_001309218.1 | 4 | 875 | 120 | ||
NM_001322293.1 | NP_001309222.1 | 3 | 764 | 120 | ||
XM_005249082.3 | XP_005249139.1 | 3 | 581 | 120 | ||
XM_006715086.2 | XP_006715149.1 | 3 | 522 | 120 | ||
XM_011514569.1 | XP_011512871.1 | 3 | 504 | 120 | ||
GenBank mRNA | AK294965.1 | - | 3 | 755 | 120 | |
U80232.1 | - | 1 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
