Assay Details
| Gene Symbol: PTP4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003463.4 | NP_003454.1 | 4 | 1428 | 77 | |
| XM_011536111.1 | XP_011534413.1 | 4 | 767 | 77 | ||
| XM_011536112.1 | XP_011534414.1 | 4 | 1526 | 77 | ||
| XM_017011270.1 | XP_016866759.1 | 4 | 784 | 77 | ||
| XM_017011271.1 | XP_016866760.1 | 4 | 764 | 77 | ||
| XM_017011272.1 | XP_016866761.1 | 4 | 1428 | 77 | ||
| GenBank mRNA | AB451314.1 | - | 3 | 275 | 77 | |
| AB451444.1 | - | 3 | 275 | 77 | ||
| AK312526.1 | - | 4 | 912 | 77 | ||
| BC023975.2 | - | 4 | 870 | 2960001 | 77 | |
| BC045571.1 | - | 4 | 1428 | 4826233 | 77 | |
| CR749458.1 | - | 6 | 1262 | 77 | ||
| U48296.1 | - | 3 | 667 | 77 | ||
| U69701.1 | - | 1 | 39 | 77 | ||
More Information
Set Membership: |
|

PTP4A1
Genomic Map