Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173685.2 | NP_775956.1 | 5 | 562 | 146 | |
| XM_005250875.2 | XP_005250932.1 | 5 | 570 | 146 | ||
| XM_005250876.4 | XP_005250933.1 | 4 | 463 | 146 | ||
| XM_011516974.2 | XP_011515276.1 | 6 | 632 | 146 | ||
| XM_011516975.1 | XP_011515277.1 | 5 | 527 | 146 | ||
| XM_017013330.1 | XP_016868819.1 | 3 | 743 | 146 | ||
| XM_017013331.1 | XP_016868820.1 | 6 | 600 | 146 | ||
| XM_017013332.1 | XP_016868821.1 | 3 | 761 | 146 | ||
| XM_017013333.1 | XP_016868822.1 | 6 | 672 | 146 | ||
| XM_017013334.1 | XP_016868823.1 | 3 | 783 | 146 | ||
| GenBank mRNA | AK057002.1 | - | 5 | 146 | ||
| BC032797.1 | - | 5 | 561 | 5109691 | 146 | |
More Information
Set Membership: |
|

NSMCE2
Genomic Map