Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000900.3 | NP_000891.2 | 4 | 376 | 76 | |
NM_001190839.1 | NP_001177768.1 | 5 | 451 | 76 | ||
GenBank mRNA | AK312029.1 | - | 4 | 311 | 76 | |
AW997681.1 | - | 1 | 215 | 76 | ||
AY542304.1 | - | 4 | 259 | 76 | ||
BC005272.1 | - | 4 | 311 | 3930143 | 76 | |
BC070314.1 | - | 4 | 284 | 6653229 | 76 | |
BC093078.1 | - | 4 | 311 | 30405585 | 76 | |
BF699158.1 | - | 390 | 76 | |||
BT006733.1 | - | 4 | 248 | 76 | ||
CB069816.1 | - | 2 | 103 | 76 | ||
CB108937.1 | - | 4 | 343 | 76 | ||
CR450358.1 | - | 4 | 248 | 76 | ||
M58549.1 | - | 4 | 274 | 76 | ||
X07362.1 | - | 4 | 254 | 76 | ||
X53331.1 | - | 4 | 294 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
