Assay Details
Gene Symbol: BCL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114735.1 | NP_001108207.1 | 1 | 573 | 96 | |
NM_004049.3 | NP_004040.1 | 1 | 573 | 96 | ||
GenBank mRNA | AL110097.1 | - | 1 | 441 | 96 | |
AY234180.1 | - | 1 | 399 | 96 | ||
BC016281.1 | - | 1 | 451 | 3920808 | 96 | |
BG198875.1 | - | 1 | 97 | |||
BG216703.1 | - | 1 | 574 | 96 | ||
BT007103.1 | - | 1 | 391 | 96 | ||
CD640106.1 | - | 1 | 339 | 96 | ||
CR541937.1 | - | 1 | 391 | 96 | ||
CR541962.1 | - | 1 | 391 | 96 | ||
U27467.1 | - | 1 | 425 | 96 | ||
U29680.1 | - | 1 | 469 | 96 | ||
Y09397.1 | - | 1 | 493 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
