Assay Details
| Gene Symbol: TUBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197181.1 | NP_001184110.1 | 3 | 409 | 57 | |
| NM_006086.3 | NP_006077.2 | 3 | 352 | 57 | ||
| GenBank mRNA | AF427491.3 | - | 3 | 229 | 57 | |
| AK058140.1 | - | 3 | 286 | 57 | ||
| AK122757.1 | - | 2 | 2311 | 57 | ||
| AK127418.1 | - | 407 | 57 | |||
| AK223039.1 | - | 3 | 286 | 57 | ||
| AK292219.1 | - | 3 | 409 | 57 | ||
| AK307201.1 | - | 4 | 425 | 57 | ||
| AK314616.1 | - | 3 | 291 | 57 | ||
| AU142509.1 | - | 3 | 289 | 57 | ||
| BC000748.2 | - | 3 | 278 | 2823044 | 57 | |
| BC001678.2 | - | 2 | 97 | 57 | ||
| BC003021.1 | - | 3 | 278 | 2823044 | 57 | |
| BC047518.1 | - | 1 | 42 | 57 | ||
| CD621827.1 | - | 3 | 306 | 57 | ||
| DA054466.1 | - | 3 | 352 | 57 | ||
| S62643.1 | - | 3 | 208 | 57 | ||
| U47634.1 | - | 3 | 229 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK122757.1, AK292219.1, NM_001197181.1 transcripts.
Set Membership: |
|

TUBB3
Genomic Map