Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130006.1 | NP_001123478.1 | 1 | 82 | 88 | |
| NM_002094.3 | NP_002085.2 | 1 | 82 | 88 | ||
| GenBank mRNA | AU132091.1 | - | 1 | 82 | 88 | |
| DC384910.1 | - | 1 | 62 | 88 | ||
| X17644.1 | - | 1 | 67 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130006.1, NM_002094.3, X17644.1 transcripts.
Set Membership: |
|

GSPT1
Genomic Map