Assay Details
| Gene Symbol: RUNDC3A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110802.1 | 4 | 495 | 84 | ||
| GenBank mRNA | AK055254.1 | - | 4 | 457 | 84 | |
| BC051705.1 | - | 3 | 252 | 84 | ||
More Information
Set Membership: |
|

RUNDC3A-AS1
Genomic Map