Assay Details
| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083908.1 | NP_001077377.1 | 1 | 184 | 59 | |
| NM_018139.2 | NP_060609.2 | 1 | 184 | 59 | ||
| GenBank mRNA | BX248765.1 | - | 1 | 184 | 59 | |
| FJ158843.1 | - | 1 | 103 | 59 | ||
More Information
Set Membership: |
|

DNAAF2
Genomic Map