Assay Details
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143947.1 | NP_001137419.1 | 7 | 1375 | 69 | |
NM_018974.3 | NP_061847.2 | 8 | 1501 | 69 | ||
XM_011535905.2 | XP_011534207.1 | 9 | 1907 | 69 | ||
XM_011535906.2 | XP_011534208.1 | 10 | 2056 | 69 | ||
XM_011535907.2 | XP_011534209.1 | 9 | 2131 | 69 | ||
XM_017010958.1 | XP_016866447.1 | 9 | 1472 | 69 | ||
GenBank mRNA | AJ422141.1 | - | 8 | 1429 | 69 | |
AJ508812.1 | - | 8 | 1403 | 69 | ||
AK091987.1 | - | 8 | 1501 | 69 | ||
BC098248.1 | - | 7 | 1226 | 40006812 | 69 | |
BC099718.1 | - | 7 | 1226 | 40006815 | 69 | |
BC105635.1 | - | 7 | 1226 | 40006813 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
