Assay Details
| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003761.4 | NP_003752.2 | 3 | 448 | 103 | |
| XM_017005170.1 | XP_016860659.1 | 4 | 537 | 103 | ||
| GenBank mRNA | AF053233.1 | - | 3 | 104 | ||
| BC001634.1 | - | 3 | 345 | 3538672 | 103 | |
| BM702226.1 | - | 3 | 448 | 103 | ||
| BQ227618.1 | - | 3 | 378 | 103 | ||
| BX092149.1 | - | 3 | 339 | 103 | ||
More Information
Set Membership: |
|

VAMP8
Genomic Map