Assay Details
| Gene Symbol: GNAS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002785.2 | 1 | 50 | 79 | ||
| GenBank mRNA | AJ251759.1 | - | 1 | 50 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNAS-AS1| Gene Symbol: GNAS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002785.2 | 1 | 50 | 79 | ||
| GenBank mRNA | AJ251759.1 | - | 1 | 50 | 79 | |
Set Membership: |
|