Assay Details
| Gene Symbol: RPL23AP64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003040.2 | 1 | 205 | 66 | ||
| GenBank mRNA | BC017930.1 | - | 1 | 202 | 66 | |
| BC060042.1 | - | 1 | 205 | 66 | ||
More Information
Set Membership: |
|

RPL23AP64
Genomic Map