Assay Details
| Gene Symbol: THPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000460.3 | NP_000451.1 | 2 - 3 | 290 | 78 | |
| NM_001177597.2 | NP_001171068.1 | 2 - 3 | 290 | 78 | ||
| NM_001177598.2 | NP_001171069.1 | 2 - 3 | 290 | 78 | ||
| NM_001289997.1 | NP_001276926.1 | 2 - 3 | 290 | 78 | ||
| NM_001289998.1 | NP_001276927.1 | 3 - 4 | 557 | 78 | ||
| NM_001290003.1 | NP_001276932.1 | 3 - 4 | 557 | 78 | ||
| NM_001290022.1 | NP_001276951.1 | 3 - 4 | 557 | 78 | ||
| NM_001290026.1 | NP_001276955.1 | 3 - 4 | 557 | 78 | ||
| NM_001290027.1 | NP_001276956.1 | 3 - 4 | 557 | 78 | ||
| NM_001290028.1 | NP_001276957.1 | 2 - 3 | 254 | 78 | ||
| XM_017007107.1 | XP_016862596.1 | 3 - 4 | 557 | 78 | ||
| GenBank mRNA | BC130322.1 | - | 2 - 3 | 222 | 40146353 | 78 |
| BC136338.1 | - | 2 - 3 | 222 | 9020325 | 78 | |
| BC143982.1 | - | 2 - 3 | 222 | 9052500 | 78 | |
| L33410.1 | - | 2 - 3 | 226 | 78 | ||
| U11025.1 | - | 1 - 2 | 46 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC130322.1, BC136338.1, BC143982.1, L33410.1, NM_000460.3, NM_001177597.2, NM_001177598.2, NM_001289997.1, NM_001289998.1, NM_001290022.1, NM_001290026.1, NM_001290027.1, NM_001290028.1, U11025.1 transcripts.
Set Membership: |
|

THPO
Genomic Map