Assay Details
| Gene Symbol: LOC730098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320037.1 | NP_001306966.1 | 1 - 2 | 80 | 72 | |
| NR_135148.1 | 1 - 2 | 80 | 72 | |||
| GenBank mRNA | BM695963.1 | - | 1 - 2 | 72 | ||
| DB061900.1 | - | 1 - 2 | 80 | 72 | ||
| HY158194.1 | - | 1 - 2 | 75 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320037.1 transcripts.
Set Membership: |
|

LOC730098
Genomic Map