Assay Details
Gene Symbol: PRORY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282471.1 | NP_001269400.1 | 1 - 2 | 128 | 114 | |
GenBank mRNA | AK026367.1 | - | 2 - 3 | 327 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK026367.1, NM_001282471.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
