Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282533.1 | NP_001269462.1 | 2 - 3 | 368 | 57 | |
| XM_005260892.1 | XP_005260949.1 | 2 - 3 | 368 | 57 | ||
| GenBank mRNA | BC038860.1 | - | 2 - 3 | 324 | 5260571 | 57 |
| HY126776.1 | - | 2 - 3 | 368 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038860.1, NM_001282533.1, XM_005260892.1 transcripts.
Set Membership: |
|

LZTS3
Genomic Map