Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282621.1 | NP_001269550.1 | 1 - 2 | 453 | 79 | |
| GenBank mRNA | AK293618.1 | - | 1 - 2 | 373 | 79 | |
More Information
Set Membership: |
|

PGRMC1
Genomic Map