Assay Details
| Gene Symbol: SLC35A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042498.2 | NP_001035963.1 | 3 - 4 | 745 | 92 | |
| NM_001282649.1 | NP_001269578.1 | 2 - 3 | 562 | 92 | ||
| NM_001282650.1 | NP_001269579.1 | 3 - 4 | 784 | 92 | ||
| NM_001282651.1 | NP_001269580.1 | 4 - 5 | 829 | 92 | ||
| NM_005660.2 | NP_005651.1 | 3 - 4 | 745 | 92 | ||
| GenBank mRNA | AK225508.1 | - | 3 - 4 | 433 | 92 | |
| AK292816.1 | - | 3 - 4 | 462 | 92 | ||
| AK293415.1 | - | 2 - 3 | 250 | 92 | ||
| AK293419.1 | - | 3 - 4 | 472 | 92 | ||
| AK298484.1 | - | 4 - 5 | 517 | 92 | ||
| CR450287.1 | - | 3 - 4 | 423 | 92 | ||
| D84454.1 | - | 3 - 4 | 746 | 92 | ||
| D88146.1 | - | 3 - 4 | 427 | 92 | ||
More Information
Set Membership: |
|

SLC35A2
Genomic Map