Assay Details
| Gene Symbol: LRIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017924.3 | NP_001017924.1 | 1 - 2 | 179 | 97 | |
| NM_001284223.1 | NP_001271152.1 | 1 - 2 | 179 | 97 | ||
| GenBank mRNA | BC140765.1 | - | 1 - 2 | 179 | 9021633 | 97 |
| BC144475.1 | - | 1 - 2 | 179 | 9053002 | 97 | |
| BX648524.1 | - | 1 - 2 | 119 | 97 | ||
More Information
Set Membership: |
|

LRIT2
Genomic Map