Assay Details
| Gene Symbol: C6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286635.1 | NP_001273564.1 | 2 - 3 | 263 | 70 | |
| NM_001286637.1 | NP_001273566.1 | 2 - 3 | 263 | 70 | ||
| XM_011514377.2 | XP_011512679.1 | 4 - 5 | 1026 | 70 | ||
| XM_017010432.1 | XP_016865921.1 | 4 - 5 | 480 | 70 | ||
| GenBank mRNA | AK300351.1 | - | 2 - 3 | 141 | 70 | |
| AK315984.1 | - | 2 - 3 | 172 | 70 | ||
| AY358637.1 | - | 2 - 3 | 137 | 70 | ||
| BC095423.1 | - | 2 - 3 | 176 | 30348810 | 70 | |
| BX647220.1 | - | 2 - 3 | 135 | 70 | ||
| FR832871.1 | - | 2 - 3 | 207 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300351.1, AK315984.1, AY358637.1, BC095423.1, FR832871.1, NM_001286635.1, NM_001286637.1, XM_011514377.2, XM_017010432.1 transcripts.
Set Membership: |
|

C6orf89
Genomic Map