Assay Details
| Gene Symbol: C6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286635.1 | NP_001273564.1 | 3 - 4 | 471 | 108 | |
| NM_152734.3 | NP_689947.2 | 2 - 3 | 371 | 108 | ||
| XM_011514377.2 | XP_011512679.1 | 5 - 6 | 1234 | 108 | ||
| XM_017010432.1 | XP_016865921.1 | 5 - 6 | 688 | 108 | ||
| XM_017010433.1 | XP_016865922.1 | 5 - 6 | 689 | 108 | ||
| XM_017010434.1 | XP_016865923.1 | 4 - 5 | 927 | 108 | ||
| GenBank mRNA | AJ420498.1 | - | 2 - 3 | 378 | 108 | |
| AK172780.1 | - | 2 - 3 | 280 | 108 | ||
| AK300351.1 | - | 3 - 4 | 349 | 108 | ||
| AY358637.1 | - | 3 - 4 | 345 | 108 | ||
| BC023605.2 | - | 2 - 3 | 229 | 108 | ||
| BC095423.1 | - | 3 - 4 | 384 | 30348810 | 108 | |
| BC105946.1 | - | 2 - 3 | 278 | 40032607 | 108 | |
| BC105947.1 | - | 2 - 3 | 279 | 40032604 | 108 | |
| BC107489.1 | - | 2 - 3 | 279 | 40032608 | 108 | |
| BX538108.1 | - | 2 - 3 | 264 | 108 | ||
| BX647220.1 | - | 3 - 4 | 343 | 108 | ||
| FR832871.1 | - | 3 - 4 | 415 | 108 | ||
| FR832872.1 | - | 2 - 3 | 400 | 108 | ||
More Information
Set Membership: |
|

C6orf89
Genomic Map