Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292016.1 | NP_001278945.1 | 3 - 4 | 742 | 89 | |
| GenBank mRNA | BX386180.2 | - | 3 - 4 | 703 | 89 | |
More Information
Set Membership: |
|

TNIP2
Genomic Map