Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302272.1 | NP_001289201.1 | 3 - 4 | 391 | 97 | |
| NM_006793.4 | NP_006784.1 | 3 - 4 | 391 | 97 | ||
| NR_126102.1 | 2 - 3 | 258 | 97 | |||
| GenBank mRNA | AK130897.1 | - | 3 - 4 | 338 | 97 | |
| AK222659.1 | - | 3 - 4 | 321 | 97 | ||
| AK313169.1 | - | 3 - 4 | 329 | 97 | ||
| AV718278.1 | - | 3 - 4 | 288 | 97 | ||
| AV761690.1 | - | 2 - 3 | 204 | 97 | ||
| BC002685.1 | - | 3 - 4 | 322 | 3607558 | 97 | |
| BC007062.1 | - | 3 - 4 | 346 | 3960644 | 97 | |
| BC008435.1 | - | 3 - 4 | 330 | 4081846 | 97 | |
| BC009601.1 | - | 3 - 4 | 323 | 4134666 | 97 | |
| BC021691.1 | - | 3 - 4 | 363 | 3961379 | 97 | |
| BC022373.1 | - | 3 - 4 | 336 | 4775243 | 97 | |
| BC059169.1 | - | 3 - 4 | 323 | 4133576 | 97 | |
| BC111397.1 | - | 3 - 4 | 330 | 4246143 | 97 | |
| BT020007.1 | - | 3 - 4 | 316 | 97 | ||
| BX382389.2 | - | 3 - 4 | 340 | 97 | ||
| CD687044.1 | - | 3 - 4 | 97 | |||
| CR450344.1 | - | 3 - 4 | 316 | 97 | ||
| D49396.1 | - | 3 - 4 | 322 | 97 | ||
| EF036492.1 | - | 2 - 3 | 291 | 97 | ||
| HY173455.1 | - | 3 - 4 | 97 | |||
More Information
Set Membership: |
|

PRDX3
Genomic Map