Assay Details
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303410.1 | NP_001290339.1 | 11 - 12 | 1292 | 127 | |
GenBank mRNA | X76060.1 | - | 11 - 12 | 1274 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
