Assay Details
| Gene Symbol: HEXIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303442.1 | NP_001290371.1 | 1 - 2 | 86 | 80 | |
| XM_011524307.2 | XP_011522609.2 | 2 - 3 | 679 | 80 | ||
| GenBank mRNA | BC025970.1 | - | 1 - 2 | 50 | 4559410 | 80 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025970.1, NM_001303442.1 transcripts.
Set Membership: |
|

HEXIM2
Genomic Map