Assay Details
| Gene Symbol: ARPP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306191.1 | NP_001293120.1 | 3 - 4 | 291 | 99 | |
| NM_001306196.1 | NP_001293125.1 | 1 - 2 | 290 | 99 | ||
| GenBank mRNA | CV802473.1 | - | 173 | 99 | ||
| HY011638.1 | - | 3 - 4 | 294 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001306191.1, NM_001306196.1 transcripts.
Set Membership: |
|

ARPP19
Genomic Map