Assay Details
| Gene Symbol: GPX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306201.1 | NP_001293130.1 | 1 - 2 | 129 | 89 | |
| GenBank mRNA | DA660205.1 | - | 1 - 2 | 65 | 89 | |
More Information
Set Membership: |
|

GPX8
Genomic Map