Assay Details
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032283.2 | NP_001027454.1 | 2 - 3 | 700 | 121 | |
NM_001032284.2 | NP_001027455.1 | 2 - 3 | 700 | 121 | ||
NM_001307975.1 | NP_001294904.1 | 2 - 3 | 700 | 121 | ||
NM_003276.2 | NP_003267.1 | 2 - 3 | 700 | 121 | ||
XM_005269132.3 | XP_005269189.1 | 2 - 3 | 691 | 121 | ||
XM_017019914.1 | XP_016875403.1 | 2 - 3 | 392 | 121 | ||
GenBank mRNA | AB209297.1 | - | 2 - 3 | 637 | 121 | |
AF070631.1 | - | 2 - 3 | 641 | 121 | ||
AK315221.1 | - | 2 - 3 | 630 | 121 | ||
BC053675.1 | - | 2 - 3 | 646 | 6170639 | 121 | |
EF028063.1 | - | 2 - 3 | 421 | 121 | ||
S76736.1 | - | 2 - 3 | 605 | 121 | ||
U09086.1 | - | 2 - 3 | 609 | 121 | ||
U09087.1 | - | 2 - 3 | 642 | 121 | ||
U09088.1 | - | 2 - 3 | 642 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019914.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
