Assay Details
| Gene Symbol: STARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308056.1 | NP_001294985.1 | 5 - 6 | 609 | 82 | |
| NM_001308059.1 | NP_001294988.1 | 6 - 7 | 644 | 82 | ||
| NM_001308060.1 | NP_001294989.1 | 3 - 4 | 374 | 82 | ||
| NM_001308061.1 | NP_001294990.1 | 4 - 5 | 501 | 82 | ||
| NM_139164.2 | NP_631903.1 | 5 - 6 | 615 | 82 | ||
| XM_017009041.1 | XP_016864530.1 | 4 - 5 | 426 | 82 | ||
| XM_017009042.1 | XP_016864531.1 | 4 - 5 | 420 | 82 | ||
| XM_017009043.1 | XP_016864532.1 | 3 - 4 | 338 | 82 | ||
| XM_017009044.1 | XP_016864533.1 | 4 - 5 | 408 | 82 | ||
| GenBank mRNA | AF480299.1 | - | 4 - 5 | 393 | 82 | |
| AK054566.1 | - | 5 - 6 | 477 | 82 | ||
| AK290312.1 | - | 5 - 6 | 528 | 82 | ||
| AK315863.1 | - | 3 - 4 | 312 | 82 | ||
| AK315869.1 | - | 4 - 5 | 426 | 82 | ||
| AL710253.1 | - | 5 - 6 | 574 | 82 | ||
| AL832599.2 | - | 1 - 2 | 71 | 82 | ||
More Information
Set Membership: |
|

STARD4
Genomic Map