Assay Details
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308226.1 | NP_001295155.1 | 1 - 2 | 156 | 64 | |
GenBank mRNA | BG705752.1 | - | 1 - 2 | 163 | 64 | |
BM558707.1 | - | 1 - 2 | 144 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001308226.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
