Assay Details
| Gene Symbol: TMIGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169126.1 | NP_001162597.1 | 3 - 4 | 492 | 55 | |
| NM_001308232.1 | NP_001295161.1 | 2 - 3 | 132 | 55 | ||
| NM_144615.2 | NP_653216.2 | 3 - 4 | 492 | 55 | ||
| XM_017026284.1 | XP_016881773.1 | 3 - 4 | 500 | 55 | ||
| GenBank mRNA | AY358964.1 | - | 3 - 4 | 454 | 55 | |
| BC015655.1 | - | 3 - 4 | 492 | 4811204 | 55 | |
| HQ452865.1 | - | 2 - 3 | 86 | 55 | ||
More Information
Set Membership: |
|

TMIGD2
Genomic Map