Assay Details
| Gene Symbol: KIAA0101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029989.2 | NP_001025160.1 | 1 - 2 | 188 | 84 | |
| NM_014736.5 | NP_055551.1 | 1 - 2 | 188 | 84 | ||
| GenBank mRNA | AF529370.1 | - | 1 - 2 | 49 | 84 | |
| AK290748.1 | - | 1 - 2 | 119 | 84 | ||
| AV757440.1 | - | 1 - 2 | 218 | 84 | ||
| AY598324.1 | - | 1 - 2 | 49 | 84 | ||
| BC005832.2 | - | 1 - 2 | 108 | 2988995 | 84 | |
| BC007101.1 | - | 1 - 2 | 115 | 4281110 | 84 | |
| BC016782.1 | - | 1 - 2 | 123 | 4109322 | 84 | |
| BF691807.1 | - | 1 - 2 | 189 | 84 | ||
| BG502841.1 | - | 181 | 84 | |||
| D14657.1 | - | 1 - 2 | 110 | 84 | ||
More Information
Set Membership: |
|

KIAA0101
Genomic Map