Assay Details
| Gene Symbol: ZNF404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033719.2 | NP_001028891.2 | 1 - 2 | 133 | 100 | |
| GenBank mRNA | AK300120.1 | - | 1 - 2 | 177 | 100 | |
| AY166785.1 | - | 1 - 2 | 77 | 100 | ||
| BC101332.1 | - | 1 - 2 | 127 | 100 | ||
| BC101333.1 | - | 1 - 2 | 133 | 40025019 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300120.1 transcripts.
Set Membership: |
|

ZNF404
Genomic Map