Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316919.1 | NP_001303848.1 | 6 - 7 | 1821 | 90 | |
NM_001316920.1 | NP_001303849.1 | 6 - 7 | 602 | 90 | ||
NM_001316921.1 | NP_001303850.1 | 6 - 7 | 891 | 90 | ||
NM_001316922.1 | NP_001303851.1 | 6 - 7 | 816 | 90 | ||
NM_001316923.1 | NP_001303852.1 | 5 - 6 | 751 | 90 | ||
NM_016078.5 | NP_057162.4 | 6 - 7 | 896 | 90 | ||
GenBank mRNA | AF151906.1 | - | 6 - 7 | 895 | 90 | |
AF223467.1 | - | 6 - 7 | 896 | 90 | ||
AK128119.1 | - | 3 - 4 | 2256 | 90 | ||
AK290813.1 | - | 6 - 7 | 620 | 90 | ||
BC008430.1 | - | 6 - 7 | 622 | 4292664 | 90 | |
BQ433266.1 | - | 5 - 6 | 517 | 90 | ||
BX537929.1 | - | 6 - 7 | 612 | 90 | ||
BX648167.1 | - | 6 - 7 | 1821 | 90 | ||
CN390782.1 | - | 5 - 6 | 594 | 90 | ||
DA773045.1 | - | 5 - 6 | 476 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
