Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316924.1 | NP_001303853.1 | 5 - 6 | 761 | 67 | |
GenBank mRNA | DA551966.1 | - | 5 - 6 | 482 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
