Assay Details
| Gene Symbol: SNRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317168.1 | NP_001304097.1 | 2 - 3 | 805 | 116 | |
| GenBank mRNA | BC035103.1 | - | 2 - 3 | 786 | 5262406 | 116 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC035103.1, NM_001317168.1 transcripts.
Set Membership: |
|

SNRPG
Genomic Map