Assay Details
| Gene Symbol: SLC10A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029998.5 | NP_001025169.1 | 1 - 2 | 353 | 92 | |
| NM_001300842.2 | NP_001287771.1 | 1 - 2 | 353 | 92 | ||
| NM_001317816.1 | NP_001304745.1 | 1 - 2 | 353 | 92 | ||
| NM_001317817.1 | NP_001304746.1 | 1 - 2 | 353 | 92 | ||
| NM_001317818.1 | NP_001304747.1 | 1 - 2 | 353 | 92 | ||
| NM_032128.4 | NP_115504.1 | 1 - 2 | 353 | 92 | ||
| NR_133924.1 | 1 - 2 | 353 | 92 | |||
| XM_011532311.2 | XP_011530613.1 | 1 - 2 | 353 | 92 | ||
| XM_017008689.1 | XP_016864178.1 | 1 - 2 | 353 | 92 | ||
| XM_017008690.1 | XP_016864179.1 | 1 - 2 | 353 | 92 | ||
| XM_017008691.1 | XP_016864180.1 | 1 - 2 | 353 | 92 | ||
| XM_017008692.1 | XP_016864181.1 | 1 - 2 | 733 | 92 | ||
| GenBank mRNA | AK291046.1 | - | 1 - 2 | 221 | 92 | |
| AL136728.1 | - | 1 - 2 | 281 | 92 | ||
| AY346324.1 | - | 1 - 2 | 316 | 92 | ||
| BC023288.1 | - | 1 - 2 | 304 | 92 | ||
| BC063471.1 | - | 1 - 2 | 266 | 92 | ||
| BC120970.1 | - | 1 - 2 | 212 | 92 | ||
| BC120971.1 | - | 1 - 2 | 212 | 92 | ||
| BC127626.1 | - | 1 - 2 | 212 | 92 | ||
| BC127627.1 | - | 1 - 2 | 212 | 92 | ||
| BC148252.1 | - | 1 - 2 | 212 | 92 | ||
| BC150308.1 | - | 1 - 2 | 322 | 8860437 | 92 | |
| CR933647.1 | - | 1 - 2 | 347 | 92 | ||
| CR936751.1 | - | 1 - 2 | 210 | 92 | ||
| DA590625.1 | - | 1 - 2 | 354 | 92 | ||
| DB205503.1 | - | 1 - 2 | 353 | 92 | ||
| DQ122860.1 | - | 1 - 2 | 99 | 92 | ||
| DQ122861.1 | - | 1 - 2 | 99 | 92 | ||
| DQ871036.1 | - | 1 - 2 | 99 | 92 | ||
More Information
Set Membership: |
|

SLC10A7
Genomic Map