Assay Details
| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317987.1 | NP_001304916.1 | 2 - 3 | 388 | 119 | |
| NM_014443.2 | NP_055258.1 | 2 - 3 | 364 | 119 | ||
| XM_011537622.2 | XP_011535924.1 | 3 - 4 | 687 | 119 | ||
| XM_017009346.1 | XP_016864835.1 | 2 - 3 | 881 | 119 | ||
| XM_017009347.1 | XP_016864836.1 | 4 - 5 | 595 | 119 | ||
| XM_017009348.1 | XP_016864837.1 | 3 - 4 | 478 | 119 | ||
| GenBank mRNA | AF110385.1 | - | 2 - 3 | 357 | 119 | |
| AF152098.1 | - | 2 - 3 | 355 | 119 | ||
| AF184969.1 | - | 2 - 3 | 355 | 119 | ||
| AF212311.1 | - | 2 - 3 | 364 | 119 | ||
| AF218727.1 | - | 2 - 3 | 355 | 119 | ||
| AY358931.1 | - | 2 - 3 | 355 | 119 | ||
| BC113925.1 | - | 2 - 3 | 337 | 40084832 | 119 | |
| BC113946.1 | - | 2 - 3 | 337 | 40084836 | 119 | |
| BM826445.1 | - | 2 - 3 | 388 | 119 | ||
| CR457192.1 | - | 2 - 3 | 314 | 119 | ||
More Information
Set Membership: |
|

IL17B
Genomic Map