Assay Details
| Gene Symbol: ZNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318137.1 | NP_001305066.1 | 3 - 4 | 348 | 79 | |
| NM_017715.3 | NP_060185.2 | 5 - 6 | 600 | 79 | ||
| GenBank mRNA | AF193057.1 | - | 5 - 6 | 563 | 79 | |
| AF217988.1 | - | 5 - 6 | 564 | 79 | ||
| AK297625.1 | - | 5 - 6 | 562 | 79 | ||
| CB129643.1 | - | 3 - 4 | 304 | 79 | ||
More Information
Set Membership: |
|

ZNF3
Genomic Map