Assay Details
| Gene Symbol: CRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320099.1 | NP_001307028.1 | 1 - 2 | 308 | 70 | |
| GenBank mRNA | BC037905.1 | - | 1 - 2 | 312 | 5276510 | 70 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037905.1, NM_001320099.1 transcripts.
Set Membership: |
|

CRADD
Genomic Map