Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320395.1 | NP_001307324.1 | 3 - 4 | 482 | 104 | |
| NR_135228.1 | 3 - 4 | 430 | 104 | |||
| XM_017006922.1 | XP_016862411.1 | 3 - 4 | 393 | 104 | ||
| GenBank mRNA | BC013916.1 | - | 3 - 4 | 390 | 3929908 | 104 |
| DA041324.1 | - | 3 - 4 | 338 | 104 | ||
More Information
Set Membership: |
|

PCNP
Genomic Map