Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320586.1 | NP_001307515.1 | 1 - 2 | 221 | 105 | |
| NM_001320587.1 | NP_001307516.1 | 1 - 2 | 221 | 105 | ||
| XM_017005406.1 | XP_016860895.1 | 1 - 2 | 486 | 105 | ||
| XM_017005407.1 | XP_016860896.1 | 1 - 2 | 486 | 105 | ||
| XM_017005409.1 | XP_016860898.1 | 1 - 2 | 486 | 105 | ||
| XM_017005411.1 | XP_016860900.1 | 1 - 2 | 486 | 105 | ||
| XM_017005412.1 | XP_016860901.1 | 1 - 2 | 486 | 105 | ||
| XM_017005413.1 | XP_016860902.1 | 1 - 2 | 486 | 105 | ||
| GenBank mRNA | BG741807.1 | - | 1 - 2 | 215 | 105 | |
| BU543943.1 | - | 1 - 2 | 221 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320586.1, NM_001320587.1, XM_017005406.1, XM_017005407.1, XM_017005409.1, XM_017005411.1, XM_017005412.1, XM_017005413.1 transcripts.
Set Membership: |
|

ACYP2
Genomic Map