Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320586.1 | NP_001307515.1 | 2 - 3 | 317 | 111 | |
| XM_017005407.1 | XP_016860896.1 | 2 - 3 | 582 | 111 | ||
| GenBank mRNA | BU543943.1 | - | 2 - 3 | 317 | 111 | |
More Information
Set Membership: |
|

ACYP2
Genomic Map