Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320586.1 | NP_001307515.1 | 5 - 6 | 554 | 112 | |
| NM_001320587.1 | NP_001307516.1 | 4 - 5 | 461 | 112 | ||
| XM_017005406.1 | XP_016860895.1 | 6 - 7 | 900 | 112 | ||
| XM_017005407.1 | XP_016860896.1 | 5 - 6 | 819 | 112 | ||
| XM_017005408.1 | XP_016860897.1 | 4 - 5 | 1528 | 112 | ||
| XM_017005409.1 | XP_016860898.1 | 4 - 5 | 726 | 112 | ||
| XM_017005411.1 | XP_016860900.1 | 6 - 7 | 900 | 112 | ||
| XM_017005412.1 | XP_016860901.1 | 6 - 7 | 900 | 112 | ||
| XM_017005413.1 | XP_016860902.1 | 6 - 7 | 900 | 112 | ||
| GenBank mRNA | BG741807.1 | - | 4 - 5 | 455 | 112 | |
| BU543943.1 | - | 5 - 6 | 554 | 112 | ||
More Information
Set Membership: |
|

ACYP2
Genomic Map